Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INTS12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142471.1 | NP_001135943.1 | 3 | 269 | ||
NM_020395.3 | NP_065128.2 | 4 | 431 | |||
XM_005263148.4 | XP_005263205.1 | 3 | 924 | |||
XM_011532143.2 | XP_011530445.1 | 4 | 1080 | |||
XM_011532145.2 | XP_011530447.1 | 5 | 567 | |||
GenBank mRNA | AF242524.1 | - | 3 | 263 | ||
AK314939.1 | - | 3 | 269 | |||
BC014442.1 | - | 4 | 379 | 4866321 |