Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1191 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079684.2 | NP_001073152.1 | 5 | 651 | ||
NM_001079685.2 | NP_001073153.1 | 5 | 630 | |||
NM_001287335.1 | NP_001274264.1 | 3 | 364 | |||
NM_001287336.1 | NP_001274265.1 | 4 | 543 | |||
NM_020444.4 | NP_065177.2 | 6 | 738 | |||
NR_109796.1 | 2 | 232 | ||||
NR_109798.1 | 3 | 345 | ||||
XM_005265941.1 | XP_005265998.1 | 5 | 551 | |||
XM_005265945.1 | XP_005266002.1 | 3 | 226 | |||
XM_005265946.4 | XP_005266003.1 | 5 | 646 | |||
XM_011534595.1 | XP_011532897.1 | 5 | 513 | |||
XM_011534596.1 | XP_011532898.1 | 4 | 462 | |||
XM_017009651.1 | XP_016865140.1 | 4 | 447 | |||
XM_017009653.1 | XP_016865142.1 | 3 | 319 | |||
GenBank mRNA | AK315428.1 | - | 6 | 695 | ||
AL390217.1 | - | 5 | 609 | |||
BC010448.1 | - | 6 | 673 | 4155679 | ||
BC017097.1 | - | 6 | 695 | 3529175 | ||
BX648859.1 | - | 6 | 737 | |||
CN341192.1 | - | 3 | 314 | |||
CR457331.1 | - | 4 | 365 | |||
DQ073392.1 | - | 4 | 365 | |||
HY114499.1 | - | 3 | 322 | |||
HY120613.1 | - | 2 | 219 |