Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYRM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020466.4 | NP_065199.1 | 3 | 392 | ||
NR_028493.1 | 3 | 551 | ||||
NR_028494.1 | 3 | 534 | ||||
NR_028495.1 | 3 | 402 | ||||
GenBank mRNA | AK024783.1 | - | 3 | 390 | ||
AK094994.1 | - | 3 | 460 | |||
AK311180.1 | - | 3 | 451 | |||
AV710669.1 | - | 3 | 392 | |||
BC009782.1 | - | 3 | 386 | 4092710 | ||
BC104668.1 | - | 3 | 375 | 6717725 |