Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRCH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243963.1 | NP_001230892.1 | 9 | 1245 | ||
NM_020871.3 | NP_065922.3 | 9 | 1245 | |||
XM_006724724.3 | XP_006724787.2 | 9 | 1227 | |||
XM_017029696.1 | XP_016885185.1 | 9 | 1254 | |||
XM_017029697.1 | XP_016885186.1 | 9 | 1255 | |||
GenBank mRNA | AB040928.1 | - | 5 | 426 | ||
BC125224.1 | - | 9 | 1238 | 40124466 | ||
BX647193.1 | - | 9 | 1245 |