Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021942.5 | NP_068761.4 | 2 | 346 | ||
NM_199053.2 | NP_951008.1 | 2 | 346 | |||
XM_017008537.1 | XP_016864026.1 | 2 | 346 | |||
XM_017008538.1 | XP_016864027.1 | 2 | 346 | |||
XM_017008539.1 | XP_016864028.1 | 1 | 91 | |||
GenBank mRNA | AK096345.1 | - | 2 | 320 | ||
AL833451.1 | - | 1 | 154 | |||
AL833571.1 | - | 2 | 325 | |||
BC051724.1 | - | 2 | 290 | 5265781 | ||
BC139745.1 | - | 2 | 237 | |||
BX538044.1 | - | 2 | 300 | |||
BX647127.1 | - | 2 | 317 | |||
BX648499.1 | - | 2 | 321 | |||
DB096222.1 | - | 2 | 346 |