Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003180.2 | NP_003171.2 | 3, 4 | 541 | ||
XM_006723339.3 | XP_006723402.1 | 3, 4 | 376 | |||
XM_006723340.3 | XP_006723403.1 | 3, 4 | 630 | |||
XM_017027175.1 | XP_016882664.1 | 3, 4 | 438 | |||
XM_017027176.1 | XP_016882665.1 | 3, 4 | 429 | |||
GenBank mRNA | AK125182.1 | - | 2, 3 | 1329 | ||
AK313067.1 | - | 3, 4 | 330 | |||
BC029962.1 | - | 3, 4 | 469 | |||
BC033683.1 | - | 3, 4 | 348 | |||
BC046157.1 | - | 3, 4 | 467 | 5768093 | ||
BQ417474.1 | - | 3, 4 | 454 | |||
DA529870.1 | - | 3, 4 | 541 | |||
DQ090951.1 | - | 2, 3 | 260 |