Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022481.5 | NP_071926.4 | 33 | 4912 | ||
XM_005268497.1 | XP_005268554.1 | 32 | 4837 | |||
XM_005268498.1 | XP_005268555.1 | 33 | 4816 | |||
XM_005268499.1 | XP_005268556.1 | 33 | 4850 | |||
XM_006714792.1 | XP_006714855.1 | 32 | 4663 | |||
XM_011537677.2 | XP_011535979.1 | 31 | 4331 | |||
GenBank mRNA | AJ310567.1 | - | 32 | 4862 | ||
AK001579.1 | - | 16 | 2836 | |||
AK024718.1 | - | 3 | 798 | |||
AK091385.1 | - | 32 | 4578 | |||
AK290052.1 | - | 33 | 4905 | |||
BC009968.2 | - | 7 | 1163 |