Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RMND5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288794.1 | NP_001275723.1 | 10, 11 | 1348 | ||
NM_001288795.1 | NP_001275724.1 | 9, 10 | 1197 | |||
NM_022762.4 | NP_073599.2 | 9, 10 | 1208 | |||
XM_005265969.3 | XP_005266026.1 | 8, 9 | 1028 | |||
XM_005265971.4 | XP_005266028.1 | 9, 10 | 1775 | |||
XM_005265972.3 | XP_005266029.1 | 8, 9 | 1155 | |||
XM_017009727.1 | XP_016865216.1 | 8, 9 | 1819 | |||
XM_017009728.1 | XP_016865217.1 | 8, 9 | 1486 | |||
GenBank mRNA | AK025971.1 | - | 6, 7 | 753 | ||
AK093481.1 | - | 9, 10 | 1149 | |||
AK093622.1 | - | 1, 2 | 1665 | |||
AK094065.1 | - | 6, 7 | 1355 | |||
AK122692.1 | - | 4, 5 | 2071 | |||
AK125337.1 | - | 10, 11 | 1295 | |||
AL137381.1 | - | 3, 4 | 321 | |||
AL713670.1 | - | 8, 9 | 1071 | |||
AY359092.1 | - | 6, 7 | 2066 | |||
BC009911.2 | - | 9, 10 | 1142 | 2820100 | ||
DQ494789.1 | - | 9, 10 | 1142 |