Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YTHDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022828.3 | NP_073739.3 | 1 | 321 | ||
XM_011543581.1 | XP_011541883.1 | 1 | 332 | |||
XM_011543582.2 | XP_011541884.1 | 1 | 306 | |||
XM_011543586.2 | XP_011541888.1 | 1 | 306 | |||
XM_017009730.1 | XP_016865219.1 | 1 | 332 | |||
XM_017009731.1 | XP_016865220.1 | 1 | 304 | |||
XM_017009732.1 | XP_016865221.1 | 1 | 304 | |||
XM_017009733.1 | XP_016865222.1 | 1 | 332 | |||
XM_017009734.1 | XP_016865223.1 | 1 | 332 | |||
XM_017009735.1 | XP_016865224.1 | 1 | 300 | |||
GenBank mRNA | AK096486.1 | - | 1 | 321 | ||
AK310855.1 | - | 1 | 306 | |||
BC137285.1 | - | 1 | 271 | 9021282 | ||
CA943109.1 | - | 1 | 305 |