Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMCX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168478.1 | NP_001161950.1 | 4 | 1462 | ||
NM_001168479.1 | NP_001161951.1 | 6 | 1578 | |||
NM_001168480.1 | NP_001161952.1 | 6 | 1572 | |||
NM_001168482.1 | NP_001161954.1 | 3 | 1361 | |||
NM_001168485.1 | NP_001161957.1 | 5 | 1459 | |||
NM_022838.3 | NP_073749.2 | 3 | 1349 | |||
GenBank mRNA | AK023031.1 | - | 2 | 1326 | ||
AK096692.1 | - | 6 | 1572 | |||
BC058904.1 | - | 3 | 1304 | 5531406 | ||
BC064983.1 | - | 3 | 1617 | 2960871 | ||
CR749476.1 | - | 1 | 3314 |