Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127235.2 | NP_001120707.2 | 9, 10 | 1966 | ||
NM_001127236.2 | NP_001120708.1 | 8, 9 | 1058 | |||
NM_001203246.1 | NP_001190175.1 | 8, 9 | 2134 | |||
NM_022913.3 | NP_075064.1 | 9, 10 | 2254 | |||
XM_006714673.3 | XP_006714736.1 | 11, 12 | 2390 | |||
XM_006714674.3 | XP_006714737.1 | 10, 11 | 2369 | |||
XM_017009754.1 | XP_016865243.1 | 10, 11 | 1743 | |||
XM_017009755.1 | XP_016865244.1 | 9, 10 | 1722 | |||
XM_017009756.1 | XP_016865245.1 | 9, 10 | 1683 | |||
XM_017009757.1 | XP_016865246.1 | 10, 11 | 2330 | |||
XM_017009758.1 | XP_016865247.1 | 9, 10 | 2309 | |||
XM_017009759.1 | XP_016865248.1 | 9, 10 | 2106 | |||
GenBank mRNA | AK024807.1 | - | 3, 4 | 492 | ||
AK307505.1 | - | 7, 8 | 907 | |||
AL136844.1 | - | 8, 9 | 1011 | |||
AL161991.1 | - | 8, 9 | 2033 | |||
AY226828.1 | - | 9, 10 | 2085 | |||
BC000267.1 | - | 9, 10 | 1468 | |||
BC070132.1 | - | 5, 6 | 737 | |||
BC113004.2 | - | 8, 9 | 2003 | 40034860 |