Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DSCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024094.2 | NP_076999.2 | 7 | 960 | ||
XM_005251065.3 | XP_005251122.1 | 6 | 816 | |||
GenBank mRNA | AK054585.1 | - | 7 | 932 | ||
AK074355.1 | - | 7 | 897 | |||
AK315356.1 | - | 7 | 886 | |||
AY358866.1 | - | 7 | 912 | |||
BC001316.1 | - | 7 | 939 | 3454505 | ||
BC001531.2 | - | 7 | 898 | 3510131 |