Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNJL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308173.1 | NP_001295102.1 | 5 | 1008 | ||
NM_024565.6 | NP_078841.3 | 6 | 1152 | |||
NR_131769.1 | 6 | 1010 | ||||
XM_006714917.3 | XP_006714980.1 | 5 | 1587 | |||
XM_011534646.2 | XP_011532948.1 | 5 | 890 | |||
XM_011534647.2 | XP_011532949.1 | 3 | 942 | |||
XM_017009846.1 | XP_016865335.1 | 6 | 1235 | |||
XM_017009847.1 | XP_016865336.1 | 4 | 787 | |||
XM_017009848.1 | XP_016865337.1 | 4 | 1147 | |||
XM_017009849.1 | XP_016865338.1 | 3 | 524 | |||
GenBank mRNA | AK024228.1 | - | 1 | 127 | ||
AK131381.1 | - | 6 | 1010 | |||
AK294000.1 | - | 5 | 884 | |||
AK302823.1 | - | 5 | 1008 | |||
BC013353.2 | - | 2 | 188 | |||
BC035871.1 | - | 6 | 1118 | 5245637 |