Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HMBOX1 SIRNA Assays ›
Gene Symbol:
HMBOX1
Gene Name:
homeobox containing 1
Gene Aliases:
HNF1LA, HOT1, PBHNF
Chromosome Location:
Chr.8: 28890370 - 29064939 on Build GRCh38
UniGene ID:
Hs.563560
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
127393
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HMBOX1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001135726.2
NP_001129198.1
3
704
NM_001324382.1
NP_001311311.1
3
564
NM_001324383.1
NP_001311312.1
3
704
NM_001324384.1
NP_001311313.1
3
707
NM_001324385.1
NP_001311314.1
3
704
NM_001324386.1
NP_001311315.1
3
704
NM_001324387.1
NP_001311316.1
3
704
NM_001324388.1
NP_001311317.1
3
564
NM_001324389.1
NP_001311318.1
3
704
NM_001324390.1
NP_001311319.1
3
707
NM_001324391.1
NP_001311320.1
3
704
NM_001324392.1
NP_001311321.1
3
707
NM_001324393.1
NP_001311322.1
4
820
NM_001324394.1
NP_001311323.1
3
704
NM_001324395.1
NP_001311324.1
3
704
NM_024567.4
NP_078843.2
4
738
NR_136757.1
3
704
NR_136758.1
3
704
XM_005273635.1
XP_005273692.1
3
680
XM_005273641.4
XP_005273698.1
3
680
XM_006716398.2
XP_006716461.1
3
680
XM_011544642.2
XP_011542944.1
3
680
XM_017013823.1
XP_016869312.1
3
712
XM_017013824.1
XP_016869313.1
3
502
XM_017013825.1
XP_016869314.1
3
680
GenBank mRNA
AK025269.1
-
1
169
AK290683.1
-
3
680
AK295320.1
-
2
343
AK310959.1
-
3
670
AY522342.1
-
3
449
BC009259.2
-
1
122
BC069242.1
-
4
738
5928365
DC341559.1
-
4
571
DC358620.1
-
3
524
DQ153248.1
-
1
50
DQ269478.1
-
3
449
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
RNA-dependent DNA biosynthetic process
transcription, DNA-templated
positive regulation of telomere maintenance via telomerase
positive regulation of chromatin binding
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of telomerase activity
double-stranded telomeric DNA binding
telomerase activity
protein binding
protein complex binding
telomeric DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0