Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDNF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024574.3 | NP_078850.3 | 4 | 1497 | ||
GenBank mRNA | AK026844.1 | - | 1 | 582 | ||
AK289615.1 | - | 4 | 1405 | |||
AY358821.1 | - | 4 | 1069 | |||
BC019351.1 | - | 4 | 1393 | 3928768 |