Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARFRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134758.3 | NP_001128230.1 | 7 | 1460 | ||
NM_001267544.2 | NP_001254473.1 | 7 | 1463 | |||
NM_001267545.2 | NP_001254474.1 | 6 | 1389 | |||
NM_001267546.2 | NP_001254475.1 | 7 | 1295 | |||
NM_001267547.2 | NP_001254476.1 | 8 | 1394 | |||
NM_001267548.2 | NP_001254477.1 | 8 | 1291 | |||
NM_001267549.2 | NP_001254478.1 | 7 | 1465 | |||
NM_003224.5 | NP_003215.1 | 8 | 1389 | |||
NR_051954.2 | 8 | 1387 | ||||
NR_051955.2 | 8 | 1392 | ||||
NR_051956.2 | 8 | 1389 | ||||
NR_051957.2 | 8 | 1384 | ||||
NR_051958.2 | 7 | 1318 | ||||
XM_011528482.2 | XP_011526784.1 | 8 | 1326 | |||
XM_011528483.1 | XP_011526785.1 | 8 | 1456 | |||
XM_017027576.1 | XP_016883065.1 | 8 | 1270 | |||
GenBank mRNA | AK095962.1 | - | 7 | 1808 | ||
AL833084.2 | - | 3 | 4883 | |||
BC021023.1 | - | 8 | 1274 | |||
BC092480.1 | - | 8 | 1275 | 30531333 | ||
X91504.1 | - | 7 | 1244 |