Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024605.3 | NP_078881.3 | 16 | 1423 | ||
XM_005263215.3 | XP_005263272.1 | 16 | 1659 | |||
XM_017008602.1 | XP_016864091.1 | 17 | 1695 | |||
GenBank mRNA | AB050785.1 | - | 16 | 1423 | ||
AK123034.1 | - | 7 | 455 | |||
AK304458.1 | - | 8 | 606 | |||
AL834360.1 | - | 2 | 64 | |||
BC011920.2 | - | 12 | 953 | |||
BC047914.1 | - | 2 | 498 | |||
BC109029.2 | - | 16 | 1413 | |||
BC109030.1 | - | 16 | 1412 | |||
BC126899.1 | - | 16 | 1419 | |||
BC128055.1 | - | 16 | 1423 | 40027832 | ||
BX647727.1 | - | 10 | 6553 |