Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003225.2 | NP_003216.1 | 2, 3 | 288 | ||
GenBank mRNA | BC032811.1 | - | 2, 3 | 272 | 5186922 | |
M12075.1 | - | 2, 3 | 247 | |||
X00474.1 | - | 2, 3 | 288 | |||
X52003.1 | - | 2, 3 | 288 |