Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUP43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198887.2 | NP_942590.1 | 8 | 2145 | ||
XM_005266960.4 | XP_005267017.1 | 8 | 3375 | |||
XM_005266961.3 | XP_005267018.1 | 9 | 2195 | |||
XM_005266962.3 | XP_005267019.1 | 8 | 4415 | |||
XM_011535799.2 | XP_011534101.1 | 8 | 3457 | |||
GenBank mRNA | AK023349.1 | - | 3 | 1331 | ||
AK074311.1 | - | 2232 | ||||
BC040121.1 | - | 6 | 1790 | |||
BC046637.1 | - | 1 | 1893 | |||
BC047539.1 | - | 1 | 3054 | |||
BC065028.1 | - | 8 | 2069 | 6042215 |