Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFAND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170796.1 | NP_001164267.1 | 6 | 410 | ||
NM_001170797.1 | NP_001164268.1 | 6 | 410 | |||
NM_024699.2 | NP_078975.2 | 6 | 410 | |||
NR_033193.1 | 5 | 370 | ||||
NR_033194.1 | 5 | 370 | ||||
NR_033195.1 | 5 | 348 | ||||
NR_033196.1 | 5 | 370 | ||||
GenBank mRNA | AK024069.1 | - | 6 | 401 | ||
AK225195.1 | - | 6 | 406 | |||
AK304111.1 | - | 5 | 353 | |||
AL833030.1 | - | 2 | 938 | |||
BC022251.2 | - | 6 | 395 | 4695118 | ||
BC051857.1 | - | 5 | 352 | 6012307 | ||
BC056877.1 | - | 5 | 353 | 6140929 | ||
BG618233.1 | - | 5 | 353 | |||
BQ427843.1 | - | 6 | 410 | |||
BQ435400.1 | - | 5 | 349 | |||
CR457323.1 | - | 6 | 387 |