Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003227.3 | NP_003218.2 | 3 | 409 | ||
XM_005250553.4 | XP_005250610.1 | 4 | 715 | |||
XM_017012573.1 | XP_016868062.1 | 4 | 517 | |||
GenBank mRNA | AF067864.1 | - | 3 | 395 | ||
AK022002.1 | - | 4 | 526 | |||
BC142630.1 | - | 3 | 395 | 40147690 | ||
BC150192.1 | - | 3 | 373 | |||
DR004213.1 | - | 3 | 552 |