Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MCTP1 SIRNA Assays ›
Gene Symbol:
MCTP1
Gene Name:
multiple C2 and transmembrane domain containing 1
Gene Aliases:
-
Chromosome Location:
Chr.5: 94703690 - 95284575 on Build GRCh38
UniGene ID:
Hs.591248
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
127446
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MCTP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001002796.3
NP_001002796.1
13
1700
NM_001297777.1
NP_001284706.1
11
1562
NM_024717.5
NP_078993.4
13
1979
XM_005272082.4
XP_005272139.1
14
2106
XM_005272090.4
XP_005272147.1
11
1341
XM_006714692.3
XP_006714755.1
11
1929
XM_011543649.2
XP_011541951.1
14
2180
XM_011543650.2
XP_011541952.1
12
1157
XM_017009855.1
XP_016865344.1
12
1380
XM_017009856.1
XP_016865345.1
13
1479
XM_017009857.1
XP_016865346.1
12
1226
XM_017009858.1
XP_016865347.1
11
1302
XM_017009859.1
XP_016865348.1
13
1304
XM_017009860.1
XP_016865349.1
12
1401
XM_017009861.1
XP_016865350.1
11
1676
XM_017009862.1
XP_016865351.1
10
1674
XM_017009863.1
XP_016865352.1
10
1302
XM_017009864.1
XP_016865353.1
12
2042
XM_017009865.1
XP_016865354.1
11
1622
GenBank mRNA
AK025997.1
-
2
67
AK057694.1
-
11
1341
AK058012.1
-
12
1246
AK091330.1
-
6
703
AK297325.1
-
10
982
AY656715.1
-
13
1979
AY656716.1
-
13
1316
BC030005.2
-
13
1461
3834212
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
calcium-mediated signaling
calcium ion binding
calcium-dependent phospholipid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-69b65fdbbd-95x48:80/100.66.76.145:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline