Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTCD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031720.3 | NP_001026890.2 | 4 | 1177 | ||
NM_024751.3 | NP_079027.2 | 4 | 1098 | |||
XM_005263222.3 | XP_005263279.1 | 4 | 1365 | |||
XM_011532248.2 | XP_011530550.1 | 4 | 1321 | |||
XM_011532249.2 | XP_011530551.1 | 4 | 1183 | |||
XM_011532252.2 | XP_011530554.1 | 4 | 1555 | |||
GenBank mRNA | AK023335.1 | - | 4 | 1356 | ||
AK225563.1 | - | 4 | 1356 | |||
AK292355.1 | - | 4 | 1180 | |||
AK303547.1 | - | 2 | 444 | |||
BC032942.2 | - | 4 | 1174 | 4838672 | ||
BX648355.1 | - | 4 | 1099 |