Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039580.1 | NP_001034669.1 | 11 | 1677 | ||
XM_006714306.3 | XP_006714369.1 | 11 | 1822 | |||
XM_011532253.1 | XP_011530555.1 | 11 | 1880 | |||
XM_011532254.2 | XP_011530556.1 | 11 | 1789 | |||
XM_011532255.2 | XP_011530557.1 | 10 | 1528 | |||
XM_017008616.1 | XP_016864105.1 | 11 | 1877 | |||
XM_017008617.1 | XP_016864106.1 | 11 | 1694 | |||
XM_017008618.1 | XP_016864107.1 | 11 | 1655 | |||
GenBank mRNA | AK024730.1 | - | 8 | 1076 | ||
AK024812.1 | - | 10 | 1508 | |||
AK301075.1 | - | 11 | 1469 | |||
AY690636.1 | - | 11 | 1677 | |||
BC128540.1 | - | 11 | 1589 | |||
BC128541.1 | - | 11 | 1592 | |||
BC146864.1 | - | 11 | 1589 | 9056792 | ||
CF144347.1 | - | 2 | 114 | |||
CR749274.1 | - | 2 | 50 |