Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105533.1 | NP_001099003.1 | 3 | 720 | ||
NM_024913.4 | NP_079189.4 | 4 | 936 | |||
XM_017012649.1 | XP_016868138.1 | 3 | 1412 | |||
GenBank mRNA | AK055624.1 | - | 3 | 1060 | ||
AK289978.1 | - | 4 | 933 | |||
AL832619.2 | - | 4 | 891 | |||
AL832638.2 | - | 4 | 760 | |||
AY358220.1 | - | 3 | 512 | |||
AY780785.1 | - | 3 | 489 | |||
BC018685.1 | - | 3 | 471 | |||
BC046360.1 | - | 3 | 936 |