Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024921.3 | NP_079197.3 | 17 | 2232 | ||
XM_005262203.3 | XP_005262260.1 | 17 | 2145 | |||
GenBank mRNA | AF309774.1 | - | 17 | 2233 | ||
AK025039.1 | - | 13 | 1823 | |||
AK025080.1 | - | 9 | 1197 | |||
AK026445.1 | - | 5 | 724 | |||
AK225193.1 | - | 9 | 1197 |