Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGFB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003239.3 | NP_003230.1 | 1 | 1405 | ||
GenBank mRNA | AK313564.1 | - | 1 | 486 | ||
BC018503.1 | - | 1 | 376 | 4477175 | ||
BT007287.1 | - | 1 | 276 | |||
EF560714.1 | - | 1 | 389 | |||
J03241.1 | - | 1 | 538 | |||
X14149.1 | - | 1 | 529 |