Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFAP3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009554.3 | NP_001009554.1 | 2 | 584 | ||
NM_001301647.1 | NP_001288576.1 | 2 | 659 | |||
NM_001301648.1 | NP_001288577.1 | 3 | 846 | |||
NM_021647.7 | NP_067679.6 | 3 | 1090 | |||
XM_005263366.3 | XP_005263423.1 | 4 | 1024 | |||
XM_005263368.3 | XP_005263425.1 | 2 | 2198 | |||
XM_017008864.1 | XP_016864353.1 | 5 | 968 | |||
XM_017008865.1 | XP_016864354.1 | 5 | 4084 | |||
XM_017008866.1 | XP_016864355.1 | 5 | 1158 | |||
XM_017008867.1 | XP_016864356.1 | 5 | 964 | |||
XM_017008868.1 | XP_016864357.1 | 4 | 1239 | |||
XM_017008869.1 | XP_016864358.1 | 3 | 4071 | |||
XM_017008870.1 | XP_016864359.1 | 3 | 4031 | |||
XM_017008871.1 | XP_016864360.1 | 3 | 971 | |||
GenBank mRNA | AB014526.1 | - | 3 | 938 | ||
AF327560.1 | - | 2 | 570 | |||
AK290041.1 | - | 3 | 1000 | |||
AK308263.1 | - | 2 | 574 | |||
AY391838.1 | - | 2 | 871 | |||
BC066912.1 | - | 2 | 553 | 5268480 | ||
BM552945.1 | - | 2 | 596 | |||
DA412844.1 | - | 2 | 546 |