Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003241.3 | NP_003232.2 | 3 | 351 | ||
XM_011534042.1 | XP_011532344.1 | 4 | 500 | |||
GenBank mRNA | AK125536.1 | - | 3 | 349 | ||
AK300523.1 | - | 3 | 220 | |||
AK312936.1 | - | 3 | 349 | |||
BC007003.1 | - | 3 | 346 | 3950865 | ||
DA870110.1 | - | 3 | 351 | |||
L34840.1 | - | 3 | 322 | |||
U31905.1 | - | 3 | 322 |