Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGFBR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195683.1 | NP_001182612.1 | 3 | 709 | ||
NM_001195684.1 | NP_001182613.1 | 4 | 545 | |||
NM_003243.4 | NP_003234.2 | 3 | 709 | |||
NR_036634.1 | 4 | 806 | ||||
XM_006710867.2 | XP_006710930.1 | 3 | 428 | |||
XM_006710868.2 | XP_006710931.1 | 3 | 394 | |||
GenBank mRNA | AJ251961.1 | - | 2 | 256 | ||
AK225799.1 | - | 3 | 487 | |||
AK291345.1 | - | 3 | 350 | |||
BC099914.1 | - | 2 | 711 | |||
BC126116.1 | - | 3 | 362 | 8991832 | ||
BC136295.1 | - | 3 | 362 | 9020282 | ||
BC143869.1 | - | 4 | 459 | |||
DB224305.1 | - | 3 | 400 | |||
DR002506.1 | - | 3 | 540 | |||
HG764816.1 | - | 4 | 545 | |||
L07594.1 | - | 4 | 542 |