Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003245.3 | NP_003236.3 | 2 | 208 | ||
GenBank mRNA | AK290324.1 | - | 2 | 172 | ||
AK291351.1 | - | 2 | 172 | |||
AK298637.1 | - | 2 | 174 | |||
AK299447.1 | - | 2 | 174 | |||
AK308249.1 | - | 2 | 174 | |||
AK315236.1 | - | 2 | 172 | |||
AL602787.1 | - | 2 | 174 | |||
BC109075.1 | - | 2 | 134 | 40034239 | ||
BC109076.1 | - | 2 | 134 | 40034240 | ||
DC373633.1 | - | 2 | 208 | |||
L10386.1 | - | 2 | 152 |