Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFHC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025184.3 | NP_079460.2 | 5 | 833 | ||
XM_006724562.2 | XP_006724625.1 | 4 | 546 | |||
GenBank mRNA | AK026496.1 | - | 5 | 686 | ||
AK291187.1 | - | 5 | 974 | |||
AK292967.1 | - | 5 | 817 | |||
AK297714.1 | - | 5 | 556 | |||
BC031039.1 | - | 5 | 828 | 5272398 | ||
CR749363.1 | - | 1 | 102 |