Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BHLHB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142524.1 | NP_001135996.1 | 4 | 1188 | ||
NM_001142525.1 | NP_001135997.1 | 4 | 1184 | |||
NM_001142526.1 | NP_001135998.1 | 4 | 1120 | |||
NM_001142527.1 | NP_001135999.1 | 4 | 1172 | |||
NM_001142528.1 | NP_001136000.1 | 4 | 1104 | |||
NM_001142529.1 | NP_001136001.1 | 3 | 1137 | |||
NM_001142530.1 | NP_001136002.1 | 2 | 1042 | |||
NM_030639.2 | NP_085142.1 | 3 | 1089 | |||
GenBank mRNA | AB051488.1 | - | 2 | 932 | ||
AF547055.1 | - | 1 | 552 | |||
BC041409.2 | - | 3 | 1077 | 5284828 |