Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SETD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306199.1 | NP_001293128.1 | 3 | 894 | ||
NM_001306200.1 | NP_001293129.1 | 3 | 894 | |||
NM_030648.3 | NP_085151.1 | 3 | 894 | |||
NR_131339.1 | 4 | 995 | ||||
GenBank mRNA | AB051504.1 | - | 3 | 529 | ||
AF448510.1 | - | 3 | 529 | |||
AF462150.1 | - | 3 | 262 | |||
AK310856.1 | - | 3 | 363 | |||
BC066361.1 | - | 4 | 434 | |||
BC121055.1 | - | 3 | 438 | 40118363 | ||
BC121056.1 | - | 3 | 438 | 40118365 |