Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VOPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284282.1 | NP_001271211.1 | 5 | 2018 | ||
NM_001284283.1 | NP_001271212.1 | 5 | 1952 | |||
NM_001284284.1 | NP_001271213.1 | 5 | 2113 | |||
NM_001321242.1 | NP_001308171.1 | 5 | 2033 | |||
NM_001321243.1 | NP_001308172.1 | 6 | 2186 | |||
NM_001321244.1 | NP_001308173.1 | 6 | 2219 | |||
NM_001321246.1 | NP_001308175.1 | 4 | 2069 | |||
NM_001321247.1 | NP_001308176.1 | 4 | 1953 | |||
NM_001321248.1 | NP_001308177.1 | 4 | 1896 | |||
NM_001321249.1 | NP_001308178.1 | 5 | 2035 | |||
NM_001321250.1 | NP_001308179.1 | 5 | 2057 | |||
NM_001321251.1 | NP_001308180.1 | 5 | 2043 | |||
NM_030796.4 | NP_110423.3 | 5 | 2147 | |||
GenBank mRNA | AB097004.1 | - | 5 | 2006 | ||
AF395824.1 | - | 4 | 1912 | |||
AK000539.1 | - | 1 | 307 | |||
AK075391.1 | - | 6 | 2081 | |||
AK090825.1 | - | 5 | 2057 | |||
AK092992.1 | - | 5 | 2091 | |||
AK126848.1 | - | 1 | 2448 | |||
AL136610.1 | - | 1 | 1960 | |||
BC016650.1 | - | 1 | 2039 | 4342536 |