Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YIPF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024947.3 | NP_001020118.1 | 6 | 1308 | ||
NM_001271732.1 | NP_001258661.1 | 5 | 1188 | |||
NM_030799.8 | NP_110426.4 | 6 | 1057 | |||
GenBank mRNA | AB014733.1 | - | 2 | 328 | ||
AF318329.1 | - | 6 | 946 | |||
AK054576.1 | - | 6 | 994 | |||
AK124065.1 | - | 6 | 1215 | |||
AK223515.1 | - | 6 | 920 | |||
AK307251.1 | - | 6 | 676 | |||
AY037152.1 | - | 6 | 989 | |||
AY358863.1 | - | 6 | 941 | |||
AY640925.1 | - | 6 | 973 | |||
AY640926.1 | - | 6 | 973 | |||
AY640927.1 | - | 6 | 973 | |||
AY640928.1 | - | 6 | 1224 | |||
AY640929.1 | - | 5 | 1104 | |||
BC007829.2 | - | 6 | 975 | 3632787 | ||
BC014253.2 | - | 6 | 995 | 4635110 | ||
BC024737.1 | - | 6 | 992 | 5109854 | ||
BX647859.1 | - | 6 | 997 | |||
CR749463.1 | - | 6 | 996 |