Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160223.1 | NP_001153695.1 | 6 | 886 | ||
NM_001160225.1 | NP_001153697.1 | 6 | 535 | |||
NM_030954.3 | NP_112216.3 | 6 | 656 | |||
NR_027668.1 | 5 | 812 | ||||
NR_027669.1 | 5 | 582 | ||||
XM_006716404.2 | XP_006716467.1 | 7 | 643 | |||
XM_006716405.2 | XP_006716468.1 | 4 | 402 | |||
XM_011544666.2 | XP_011542968.1 | 6 | 440 | |||
XM_017013881.1 | XP_016869370.1 | 7 | 583 | |||
XM_017013882.1 | XP_016869371.1 | 5 | 428 | |||
GenBank mRNA | AK027748.1 | - | 6 | 572 | ||
AK090864.1 | - | 5 | 745 | |||
AK225086.1 | - | 6 | 536 | |||
AK225568.1 | - | 6 | 552 | |||
AL136620.1 | - | 6 | 540 | |||
BC013422.1 | - | 5 | 447 | |||
BC039461.1 | - | 6 | 643 | |||
BC044566.1 | - | 6 | 656 | |||
BC058289.1 | - | 6 | 519 |