Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TATDN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146160.1 | NP_001139632.1 | 3 | 247 | ||
NM_001317889.1 | NP_001304818.1 | 5 | 363 | |||
NM_001317890.1 | NP_001304819.1 | 3 | 346 | |||
NM_032026.3 | NP_114415.1 | 5 | 363 | |||
NR_027427.1 | 5 | 363 | ||||
XM_006716666.3 | XP_006716729.1 | 5 | 383 | |||
XM_006716667.3 | XP_006716730.1 | 5 | 428 | |||
XM_006716668.3 | XP_006716731.1 | 5 | 441 | |||
XM_006716669.3 | XP_006716732.1 | 3 | 325 | |||
XM_006716670.3 | XP_006716733.1 | 4 | 284 | |||
XM_006716671.3 | XP_006716734.1 | 4 | 362 | |||
XM_011517331.2 | XP_011515633.1 | 4 | 279 | |||
XM_017013895.1 | XP_016869384.1 | 5 | 373 | |||
XM_017013896.1 | XP_016869385.1 | 5 | 428 | |||
XM_017013897.1 | XP_016869386.1 | 4 | 349 | |||
XM_017013898.1 | XP_016869387.1 | 4 | 286 | |||
XM_017013899.1 | XP_016869388.1 | 5 | 359 | |||
GenBank mRNA | AF212250.1 | - | 5 | 346 | ||
AK312204.1 | - | 5 | 359 | |||
AY071865.1 | - | 3 | 236 | |||
BC064964.1 | - | 5 | 347 | 6141834 | ||
BG532054.1 | - | 3 | 336 | |||
BX332564.2 | - | 5 | 345 |