Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM172A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163417.1 | NP_001156889.1 | 10 | 2900 | ||
NM_001163418.1 | NP_001156890.1 | 9 | 2708 | |||
NM_032042.5 | NP_114431.2 | 11 | 3024 | |||
NR_028080.1 | 9 | 2731 | ||||
XM_005272105.3 | XP_005272162.1 | 10 | 2797 | |||
XM_005272106.3 | XP_005272163.1 | 10 | 2771 | |||
XM_005272108.3 | XP_005272165.1 | 10 | 2918 | |||
XM_005272110.3 | XP_005272167.1 | 9 | 2625 | |||
XM_006714717.3 | XP_006714780.1 | 11 | 3016 | |||
XM_006714718.3 | XP_006714781.1 | 9 | 3630 | |||
XM_006714719.3 | XP_006714782.1 | 9 | 2779 | |||
XM_011543672.2 | XP_011541974.1 | 11 | 2920 | |||
XM_011543673.2 | XP_011541975.1 | 10 | 2819 | |||
XM_017009952.1 | XP_016865441.1 | 11 | 3312 | |||
XM_017009953.1 | XP_016865442.1 | 11 | 3310 | |||
XM_017009955.1 | XP_016865444.1 | 10 | 3728 | |||
XM_017009956.1 | XP_016865445.1 | 10 | 3042 | |||
XM_017009957.1 | XP_016865446.1 | 9 | 2996 | |||
XM_017009958.1 | XP_016865447.1 | 9 | 2791 | |||
XM_017009960.1 | XP_016865449.1 | 8 | 2573 | |||
GenBank mRNA | BC020584.2 | - | 10 | 2827 | 4398813 |