Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPHK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142601.1 | NP_001136073.1 | 6 | 784 | ||
NM_001142602.1 | NP_001136074.1 | 5 | 829 | |||
NM_021972.3 | NP_068807.2 | 6 | 826 | |||
NM_182965.2 | NP_892010.2 | 6 | 1147 | |||
XM_005257766.2 | XP_005257823.1 | 6 | 1139 | |||
GenBank mRNA | AF200328.1 | - | 5 | 410 | ||
AF238083.1 | - | 6 | 728 | |||
AF266756.1 | - | 5 | 661 | |||
AJ245504.1 | - | 4 | 383 | |||
AK022402.1 | - | 6 | 784 | |||
AK023393.1 | - | 5 | 741 | |||
AK095578.1 | - | 5 | 1469 | |||
AK122781.1 | - | 8 | 1201 | |||
AK225250.1 | - | 6 | 747 | |||
AK292294.1 | - | 6 | 871 | |||
AK292614.1 | - | 5 | 745 | |||
BC004112.2 | - | 2 | 101 | |||
BC008040.2 | - | 6 | 721 | 3831657 | ||
BC009419.1 | - | 6 | 814 | 3506924 | ||
BC014439.1 | - | 6 | 1084 | 4871343 | ||
BC030553.1 | - | 6 | 1106 | 5213270 |