Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003255.4 | NP_003246.1 | 2 | 469 | ||
GenBank mRNA | AK294290.1 | - | 2 | 229 | ||
AL713764.1 | - | 1 | 254 | |||
BC039613.1 | - | 1 | 207 | |||
BC040039.1 | - | 1 | 119 | |||
BC040445.1 | - | 1 | 204 | |||
BC052605.1 | - | 2 | 446 | 6267022 | ||
BC071586.1 | - | 2 | 466 | 30337524 | ||
J05593.1 | - | 2 | 437 | |||
M32304.1 | - | 2 | 421 | |||
S48568.1 | - | 2 | 437 | |||
X54533.1 | - | 2 | 80 |