Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INMT-FAM188B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037598.1 | 9 | 1744 | |||
GenBank mRNA | AK308762.1 | - | 9 | 1742 |
Gene Symbol: FAM188B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032222.2 | NP_115598.2 | 7 | 1292 | ||
GenBank mRNA | AF538962.1 | - | 7 | 1285 | ||
AK026027.1 | - | 7 | 1292 | |||
BC098569.1 | - | 7 | 1239 | 30924492 |