Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017698.2 | NP_060168.2 | 4 | 296 | ||
NM_032227.3 | NP_115603.2 | 5 | 887 | |||
XM_005262205.4 | XP_005262262.1 | 5 | 913 | |||
XM_005262206.3 | XP_005262263.1 | 5 | 1182 | |||
XM_005262208.4 | XP_005262265.1 | 5 | 342 | |||
XM_011531051.2 | XP_011529353.1 | 5 | 571 | |||
XM_011531054.1 | XP_011529356.1 | 5 | 560 | |||
XM_011531055.2 | XP_011529357.1 | 3 | 240 | |||
XM_017029894.1 | XP_016885383.1 | 4 | 659 | |||
XM_017029896.1 | XP_016885385.1 | 5 | 474 | |||
XM_017029897.1 | XP_016885386.1 | 4 | 551 | |||
XM_017029898.1 | XP_016885387.1 | 4 | 1132 | |||
XM_017029899.1 | XP_016885388.1 | 6 | 635 | |||
XM_017029900.1 | XP_016885389.1 | 4 | 522 | |||
GenBank mRNA | AK026332.1 | - | 3 | 770 | ||
AK094127.1 | - | 4 | 296 | |||
BC085605.1 | - | 5 | 867 | 6142249 | ||
CN349076.1 | - | 2 | 99 |