Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017698.2 | NP_060168.2 | 6 | 687 | ||
NM_032227.3 | NP_115603.2 | 7 | 1278 | |||
XM_005262205.4 | XP_005262262.1 | 7 | 1304 | |||
XM_005262208.4 | XP_005262265.1 | 7 | 733 | |||
XM_011531051.2 | XP_011529353.1 | 7 | 962 | |||
XM_011531054.1 | XP_011529356.1 | 7 | 951 | |||
XM_011531055.2 | XP_011529357.1 | 5 | 631 | |||
XM_017029894.1 | XP_016885383.1 | 6 | 1050 | |||
XM_017029897.1 | XP_016885386.1 | 6 | 942 | |||
XM_017029899.1 | XP_016885388.1 | 8 | 1026 | |||
XM_017029900.1 | XP_016885389.1 | 6 | 913 | |||
GenBank mRNA | AK026332.1 | - | 5 | 1160 | ||
AK094127.1 | - | 6 | 687 | |||
BC085605.1 | - | 7 | 1258 | 6142249 | ||
CN349076.1 | - | 4 | 490 |