Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032290.3 | NP_115666.2 | 17 | 2290 | ||
XM_017009978.1 | XP_016865467.1 | 17 | 2399 | |||
XM_017009979.1 | XP_016865468.1 | 17 | 7052 | |||
XM_017009980.1 | XP_016865469.1 | 16 | 2322 | |||
XM_017009981.1 | XP_016865470.1 | 17 | 2250 | |||
XM_017009982.1 | XP_016865471.1 | 17 | 2471 | |||
XM_017009983.1 | XP_016865472.1 | 7 | 933 | |||
XM_017009984.1 | XP_016865473.1 | 11 | 1324 | |||
GenBank mRNA | AK299080.1 | - | 17 | 2316 | ||
AK309859.1 | - | 4 | 486 | |||
AL136560.1 | - | 3 | 322 | |||
BC063674.1 | - | 3 | 792 | 4368392 | ||
CR533539.1 | - | 3 | 302 | |||
CR936768.1 | - | 17 | 2296 |