Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032290.3 | NP_115666.2 | 19 | 2528 | ||
XM_017009978.1 | XP_016865467.1 | 19 | 2637 | |||
XM_017009979.1 | XP_016865468.1 | 19 | 7290 | |||
XM_017009980.1 | XP_016865469.1 | 18 | 2560 | |||
XM_017009981.1 | XP_016865470.1 | 19 | 2488 | |||
XM_017009982.1 | XP_016865471.1 | 19 | 2709 | |||
XM_017009983.1 | XP_016865472.1 | 9 | 1171 | |||
XM_017009984.1 | XP_016865473.1 | 13 | 1562 | |||
GenBank mRNA | AK299080.1 | - | 19 | 2554 | ||
AK309859.1 | - | 6 | 724 | |||
AL136560.1 | - | 5 | 560 | |||
BC063674.1 | - | 5 | 1030 | 4368392 | ||
CR533539.1 | - | 5 | 540 | |||
CR936768.1 | - | 19 | 2534 |