Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UTP23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032334.2 | NP_115710.2 | 1, 2 | 291 | ||
GenBank mRNA | AK055279.1 | - | 1, 2 | 240 | ||
AK315770.1 | - | 1, 2 | 235 | |||
BC005955.1 | - | 1, 2 | 291 | 4285502 | ||
BC022441.1 | - | 1, 2 | 291 | 4285036 | ||
BP231814.1 | - | 1, 2 | 291 |