Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003262.3 | NP_003253.1 | 2 | 105 | ||
XM_011513114.2 | XP_011511416.1 | 3 | 182 | |||
GenBank mRNA | BC003677.1 | - | 2 | 97 | ||
BC007040.1 | - | 2 | 97 | |||
BC012035.1 | - | 2 | 89 | 4706586 | ||
BC105971.1 | - | 188 | ||||
BX475495.1 | - | 2 | 106 | |||
BX537668.1 | - | 1 | 8068 | |||
BX648539.1 | - | 2 | 89 | |||
D87127.1 | - | 2 | 86 | |||
KC005990.1 | - | 2 | 74 | |||
U93239.1 | - | 2 | 86 |