Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GFM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281302.1 | NP_001268231.1 | 13 | 1646 | ||
NM_032380.4 | NP_115756.2 | 12 | 1528 | |||
NM_170681.2 | NP_733781.1 | 12 | 1528 | |||
NM_170691.2 | NP_733792.1 | 12 | 1528 | |||
NR_104006.1 | 12 | 1528 | ||||
XM_011543691.2 | XP_011541993.1 | 12 | 1311 | |||
XM_017009986.1 | XP_016865475.1 | 12 | 1422 | |||
GenBank mRNA | AF111808.1 | - | 12 | 1278 | ||
AF367997.1 | - | 12 | 1374 | |||
AK025314.1 | - | 12 | 1305 | |||
AK315705.1 | - | 12 | 1307 | |||
AY453587.1 | - | 12 | 1272 | |||
BC015712.1 | - | 13 | 1380 | 3922036 | ||
BC030612.1 | - | 12 | 1277 | 4811939 |