Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD14B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032558.2 | NP_115947.2 | 7 | 1056 | ||
XM_006717305.3 | XP_006717368.1 | 7 | 1031 | |||
XM_011519114.2 | XP_011517416.1 | 7 | 747 | |||
XM_017015217.1 | XP_016870706.1 | 7 | 857 | |||
XM_017015218.1 | XP_016870707.1 | 7 | 784 | |||
XM_017015219.1 | XP_016870708.1 | 6 | 594 | |||
XM_017015220.1 | XP_016870709.1 | 6 | 608 | |||
XM_017015221.1 | XP_016870710.1 | 7 | 747 | |||
GenBank mRNA | AK027659.1 | - | 3 | 361 | ||
AK222836.1 | - | 3 | 371 | |||
AK300615.1 | - | 6 | 628 | |||
BC021093.1 | - | 2 | 166 | |||
BC106062.1 | - | 7 | 793 | 6162855 | ||
BX648846.1 | - | 4 | 470 |